missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INSR Recombinant Rabbit Monoclonal Antibody (23GB2050), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | INSR |
|---|---|
| Clone | 23GB2050 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088663
![]() |
Invitrogen™
MA553208 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089302
|
Invitrogen™
MA553209 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INSR Recombinant Monoclonal antibody specifically detects INSR in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| INSR | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P06213, P15127, P15208 | |
| 16337, 24954, 3643 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2050 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4932439J01Rik; CD 220; CD220; CD220 antigen; CD220 beta; CD221; D630014A15Rik; HHF 5; HHF5; IGF-I receptor; INSR; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR 1; IR beta; IR1; IR-1; IR-A; IR-B | |
| A synthesized peptide derived from human Insulin Receptor (910-976AA). | |
| Primary | |
| -20°C | |
| INSR |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title