missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INSR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115426
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total IR.
INSR is a receptor tyrosine kinase. Binding of insulin to the extracellular domain of INSR stimulates glucose uptake. INSR and IGF-1 receptors share major structural and functional similarity. The earliest cellular response to insulin stimulation is autophosphorylation of tyrosine in INSR. In humans, the INSR gene is located on chromosome 19. Defects in INSR are the cause of various insulin resistance syndromes and IGF-1R defects may also cause some forms of growth retardation.
Specifications
| INSR | |
| Polyclonal | |
| Unconjugated | |
| INSR | |
| 4932439J01Rik; CD 220; CD220; CD220 antigen; CD220 beta; CD221; D630014A15Rik; HHF 5; HHF5; IGF-I receptor; INSR; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR 1; IR beta; IR1; IR-1; IR-A; IR-B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3643 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P06213 | |
| INSR | |
| A synthesized peptide derived from human INSR(Accession P06213), corresponding to amino acid residues L1329-S1379. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction