missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INSR beta Monoclonal Antibody (CT-3)
Mouse Monoclonal Antibody
Brand: Invitrogen™ AHR0271
This item is not returnable.
View return policy
Description
Staining of formalin-fixed paraffin embedded tissue sections requires boiling the sections in 10 mM citrate buffer, pH 6.0, for 10-20 minutes followed by cooling at room temperature for 20 minutes.
Biological actions of insulin and IGF-1 are mediated by their respective cell surface receptor tyrosine kinases that regulate multiple signaling pathways through activation of a series of phosphorylation cascades. The insulin receptor. Insulin/IGF-1 binding to the extracellular domain leads to autophosphorylation of downstream target proteins. These two receptors differ in sequence in regions that confer specificity for the designated ligand as well as in certain intracellular signaling domains, resulting in significant differences in the functional consequences of activation of each receptor. Defects in IR are the cause of various insulin resistance syndromes and IGF-1R defects may cause some forms of growth retardation. Both these signaling cascades are also important for the development of cancer.
Specifications
| INSR beta | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P06213, P15208 | |
| INSR | |
| Recombinant fragment including the C-terminal 100 amino acid residues of human insulin receptor. | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CT-3 | |
| Unconjugated | |
| INSR | |
| 4932439J01Rik; CD220; CD220 beta; CD221; D630014A15Rik; HHF5; IGF-I receptor; Insr; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR beta; IR-A; IR-B | |
| Mouse | |
| Protein G | |
| RUO | |
| 16337, 3643 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction