Learn More
Invitrogen™ INSR beta Monoclonal Antibody (CT-1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA513778
Description
MA5-13778 targets Insulin Receptor beta in ELISA, AP, FACS, ICC/IF and IP applications and shows reactivity with Human, mouse, and Rat samples. The MA5-13778 immunogen is a 15-mer synthetic peptide corresponding to aa Tyr-KKNGRILTLPRSNPS from the C-terminal of human insulin receptor beta-subunit.
Biological actions of insulin and IGF-1 are mediated by their respective cell surface receptor tyrosine kinases that regulate multiple signaling pathways through activation of a series of phosphorylation cascades. The insulin receptor. Insulin/IGF-1 binding to the extracellular domain leads to autophosphorylation of downstream target proteins. These two receptors differ in sequence in regions that confer specificity for the designated ligand as well as in certain intracellular signaling domains, resulting in significant differences in the functional consequences of activation of each receptor. Defects in IR are the cause of various insulin resistance syndromes and IGF-1R defects may cause some forms of growth retardation. Both these signaling cascades are also important for the development of cancer.
Specifications
| INSR beta | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P06213, P15127, P15208 | |
| INSR | |
| A 15-mer synthetic peptide corresponding to aa Tyr-KKNGRILTLPRSNPS from the C-terminal of human insulin receptor beta-subunit (aa 1368-1382). | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 κ |
| Affinity Purification, ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| CT-1 | |
| Unconjugated | |
| INSR | |
| 4932439J01Rik; CD220; CD220 beta; CD221; D630014A15Rik; HHF5; IGF-I receptor; Insr; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR beta; IR-A; IR-B | |
| Mouse | |
| Protein G | |
| RUO | |
| 16337, 24954, 3643 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.