Learn More
Brady™ Inspection Timer
Click to view available options
Time Range:
30 days
365 days
7 days
Unit Size:
Pack of 10
Description
- Clearly highlights when the next planned maintenance intervention is due
- Versions are available that count down 7 days, 30 days, 180 days and 365 days
- Each version is equipped with a dark green, light green, yellow and flashing red LED light that indicate a recently inspected up to an uncertain equipment status
Specifications
Specifications
| Time Range | 365 days |
| Display | LED |
| Battery Type | (3) AAA Alkaline |
| Weight (Metric) | 1030 g |