Learn More
Abeba™ Insoles 3553
Product Code. 15600138
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15600138
Brand: Abeba™ 3553/43
Only null left
Add to basket
Add to basket
ESD dissipative insoles include antibacterial and antifungal properties due to active carbon.
  • Breathable textile
  • Blue
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Areal pressure absorption, climatising, comfortable
  • Benefit for joints, musculature, ligaments and spine
Specifications
Color Blue
Material Textile
Size 43
Style 3553