missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | INSM2 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18294772
|
Novus Biologicals
NBP2-55336 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675246
|
Novus Biologicals
NBP2-55336-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INSM2 Polyclonal specifically detects INSM2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| INSM2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 84684 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AEGKENSRIERTADQHPQARDSSGADQHPDSAPRQGLQVLTHPEPPLPQGPYTEGVLGRRVPVPGSTSGGRGSEIFVCP | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| insulinoma-associated 2, Nbla106, zinc finger protein IA-6 | |
| INSM2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel