missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSM1 Antibody (INSM1/6286R), DyLight 755, Novus Biologicals™
Description
INSM1 Monoclonal antibody specifically detects INSM1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | INSM1 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | INSM1/6286R |
| Conjugate | DyLight 755 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | IA1IA-1, insulinoma-associated 1, insulinoma-associated protein 1, Zinc finger protein IA-1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fragment (around aa81-125) corresponding to the N-terminus of human INSM1 (Uniprot: Q01101 ) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?