missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INSL5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103442
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total INSL5.
The protein encoded by this gene contains a classical signature of the insulin superfamily and is highly similar to relaxin 3 (RLN3/INSL7).
Specifications
| INSL5 | |
| Polyclonal | |
| Unconjugated | |
| INSL5 | |
| Insl5; insulin like 5; insulin-like 5; insulin-like peptide 5; Insulin-like peptide INSL5; Insulin-like peptide INSL5 A chain; Insulin-like peptide INSL5 B chain; prepro-INSL5; PRO182; Relaxin/insulin-like factor 2; relaxin/insulin-like protein; Rif; Rif2; UNQ156; UNQ156/PRO182; Zins3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10022, 102554482, 23919 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9WUG6, Q9Y5Q6 | |
| INSL5 | |
| A synthesized peptide derived from human INSL5(Accession Q9Y5Q6), corresponding to amino acid residues L31-N81. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction