missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£166.00 - £366.00
Specifications
| Antigen | INSL3 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18320767
|
Novus Biologicals
NBP3-04435-20UL |
20 μL |
£166.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18383474
|
Novus Biologicals
NBP3-04435-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INSL3 Polyclonal specifically detects INSL3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| INSL3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 3640 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| insulin-like 3, insulin-like 3 (Leydig cell), leydig insulin -like hormone, leydig insulin -like peptide, Leydig insulin-like peptide, Ley-I-L, MGC119818, MGC119819, Relaxin-like factor, relaxin-like factor b, RLFley-I-L, RLNL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-131 of human INSL3 (NP_005534.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title