missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INSIG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576887
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
INSIG2 is highly similar to the protein product encoded by gene INSIG1. Both INSIG1 protein and this protein are endoplasmic reticulum proteins that block the processing of sterol regulatory element binding proteins (SREBPs) by binding to SREBP cleavage-activating protein (SCAP), and thus prevent SCAP from escorting SREBPs to the Golgi.The protein encoded by this gene is highly similar to the protein product encoded by gene INSIG1. Both INSIG1 protein and this protein are endoplasmic reticulum proteins that block the processing of sterol regulatory element binding proteins (SREBPs) by binding to SREBP cleavage-activating protein (SCAP), and thus prevent SCAP from escorting SREBPs to the Golgi.
Specifications
| INSIG2 | |
| Polyclonal | |
| Unconjugated | |
| INSIG2 | |
| 2900053I11Rik; C730043J18Rik; Insig2; INSIG-2; INSIG2 membrane protein; INSIG-2 membrane protein; insulin induced factor 2; insulin induced gene 2; insulin induced protein 2; insulin-induced 2 protein; insulin-induced gene 2 protein; insulin-induced membrane protein 2; MGC26273 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51141, 72999 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q91WG1, Q9Y5U4 | |
| INSIG2 | |
| KLH conjugated synthetic peptide derived from human Insig2: 65-160. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction