missing translation for 'onlineSavingsMsg'
Learn More

INSIG-1 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Product Code. 18318676 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18318676 20 μL 20µL
18327486 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18318676 Supplier Novus Biologicals Supplier No. NBP30296920UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

INSIG-1 Polyclonal specifically detects INSIG-1 in Human, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INSIG-1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias CL6, CL-6, INSIG-1, INSIG-1 membrane protein, insulin induced gene 1, insulin-induced gene 1 protein, MGC1405
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human INSIG-1 (NP_005533.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, ER Markers, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3638
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.