missing translation for 'onlineSavingsMsg'
Learn More

INSIG-1 Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

£171.00 - £401.00

Specifications

Antigen INSIG-1
Dilution ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30232984
View Documents
Novus Biologicals
NBP3-33431-100ul
100 μL
£401.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30228787
View Documents
Novus Biologicals
NBP3-33431-20ul
20 μL
£171.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

INSIG-1 Monoclonal antibody specifically detects INSIG-1 in Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications

Specifications

INSIG-1
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cholesterol Metabolism, ER Markers, Lipid and Metabolism, Signal Transduction
PBS (pH 7.3), 50% glycerol, 0.05% BSA
3638
IgG
Affinity purified
ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Monoclonal
Purified
RUO
Mouse, Rat
CL6, CL-6, INSIG-1, INSIG-1 membrane protein, insulin induced gene 1, insulin-induced gene 1 protein, MGC1405
Recombinant fusion protein containing a sequence corresponding to amino acids 177-277 of human INSIG-1 (NP_005533.2).,, Sequence:, ASAKLDFANNVQLSLTLAALSLGLWWTFDRSRSGLGLGITIAFLATLITQFLVYNGVYQYTSPDFLYIRSWLPCIFFSGGVTVGNIGRQLAMGVPEKPHSD
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.