missing translation for 'onlineSavingsMsg'
Learn More

INPP5F Antibody (OTI1D4) [DyLight 350], Novus Biologicals™

Product Code. 30687965 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687965 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687965 Supplier Novus Biologicals Supplier No. NBP345038UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

INPP5F Monoclonal antibody specifically detects INPP5F in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen INPP5F
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI1D4
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3, EC 3.1.3.-, EC 3.1.3.16, EC 3.1.3.36, FLJ13081, hSac2, Inositol polyphosphate 5-phosphatase F, inositol polyphosphate-5-phosphatase F, KIAA0966MGC59773, MGC131851, phosphatidylinositide phosphatase SAC2, Sac domain-containing inositol phosphatase 2, Sac domain-containing phosphoinositide 5-phosphatase 2, SAC2MSTPO47
Host Species Mouse
Immunogen Full length human recombinant protein of human INPP5F (NP_055752) produced in HEK293T cell.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22876
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.