missing translation for 'onlineSavingsMsg'
Learn More

INPP5E Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£153.00 - £366.00

Specifications

Antigen INPP5E
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18305762
View Documents
Novus Biologicals
NBP3-05081-20UL
20 μL
£153.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18306242
View Documents
Novus Biologicals
NBP3-05081-100UL
100 μL
£366.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

INPP5E Polyclonal specifically detects INPP5E in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

INPP5E
Polyclonal
Purified
RUO
Human, Rat
CORS172 kDa inositol polyphosphate 5-phosphatase, CPD4, EC 3.1.3.36, inositol polyphosphate-5-phosphatase, 72 kDa, JBTS1, Joubert syndrome 1, MGC117201, MORMS, phosphatidylinositol (4,5) bisphosphate 5-phosphatase, Phosphatidylinositol polyphosphate 5-phosphatase type IV, Phosphatidylinositol-4,5-bisphosphate 5-phosphatase, PPI5PIV
Recombinant fusion protein containing a sequence corresponding to amino acids 510-630 of human INPP5E (NP_063945.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
Rabbit
Lipid and Metabolism
PBS with 50% glycerol, pH7.3.
56623
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.