missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INPP5E Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537119
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 72 kDa. Purity is >95% by SDS-PAGE.
INPP5E (inositol polyphosphate 5-phosphatase), also known as phosphatidylinositol polyphosphate 5-phosphatase type IV, is a 644 amino acid peripheral membrane protein associated with Golgi stacks. Belonging to the inositol-1, 4, 5-trisphosphate 5-phosphatase type IV family, INPP5E converts phosphatidylinositol-3, 4, 5-triphosphate (PtdIns 3, 4, 5-P3) to PtdIns-P2. While inactive towards water soluble inositol phosphates, the activity of INPP5E is specific for lipid substrates. INPP5E becomes phosphorylated upon DNA damage and is expressed in brain, heart, pancreas, testis and spleen. Defects in INPP5E are the cause of Joubert syndrome type 1 (JBTS1), a disorder presenting with cerebellar ataxia, oculomotor apraxia, hypotonia, neonatal breathing abnormalities and psychomotor delay. In addition, mutations in the INPP5E gene may lead to mental retardation-truncal obesity-retinal dystrophy-micropenis (MORMS), an autosomal recessive disorder characterized by moderate mental retardation, truncal obesity, congenital non-progressive retinal dystrophy, and micropenis in males.
Specifications
| INPP5E | |
| Polyclonal | |
| Unconjugated | |
| Inpp5e | |
| 1200002L24Rik; 5-phosphatase that induces arborization; 72 kDa inositol polyphosphate 5-phosphatase; 72kDa; CORS1; CPD4; inositol polyphosphate 5-phosphatase; inositol polyphosphate-5-phosphatase E; inositol polyphosphate-5-phosphatase, 72 kDa; INPP5E; JBTS1; mKIAA0123; MORMS; Pharbin; phosphatidylinositol 4,5-bisphosphate 5-phosphatase; Phosphatidylinositol polyphosphate 5-phosphatase type IV; Phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase; phosphatidylinositol-4,5-bisphosphate 5-phosphatase; PPI5PIV | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114089, 56623, 64436 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide | |
| Q9JII1, Q9NRR6, Q9WVR1 | |
| Inpp5e | |
| A synthetic peptide corresponding to residues in human INPP5E. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction