missing translation for 'onlineSavingsMsg'
Learn More

INPP5A Antibody (3D8), Novus Biologicals™

Product Code. 18367788 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367788 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367788 Supplier Novus Biologicals Supplier No. H00003632M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

INPP5A Monoclonal antibody specifically detects INPP5A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen INPP5A
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 3D8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005530
Gene Alias 43 kDa inositol polyphosphate 5-phophatase, CTCL tumor antigen HD-CL-02,5PTASE, DKFZp434A1721, EC 3.1.3.56, inositol polyphosphate-5-phosphatase, 40kD, inositol polyphosphate-5-phosphatase, 40kDa, inositol trisphosphate-5-phosphatase, 40kD, InsP3 5-phosphatase, MGC116947,5PTase, MGC116949, type I inositol-14,5-trisphosphate 5-phophatase, type I inositol-14,5-trisphosphate 5-phosphatase
Host Species Mouse
Immunogen INPP5A (NP_005530, 288 a.a. ~ 387 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. YFNQEVFRDNNGTALLEFDKELSVFKDRLYELDISFPPSYPYSEDARQGEQYMNTRCPAWCDRILMSPSAKELVLRVSVCCPSPGHRGMWSAGSGLAQPW
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 3632
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.