missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INPP4B Recombinant Rabbit Monoclonal Antibody (23GB2530), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | INPP4B |
|---|---|
| Clone | 23GB2530 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089472
![]() |
Invitrogen™
MA552508 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088136
|
Invitrogen™
MA552509 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INPP4B Recombinant Monoclonal antibody specifically detects INPP4B in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| INPP4B | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O15327, Q6P1Y8, Q9QWG5 | |
| 116699, 234515, 8821 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2530 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| D-myo-inositol-3,4-bisphosphate 4-phosphohydrolase; E130107I17Rik; inositol polyphosphate 4-phosphatase II; 4-phosphatase II; inositol polyphosphate 4-phosphatase type II; inositol polyphosphate 4-phosphatase type II beta isoform; inositol polyphosphate-4-phosphatase type II 105kD; inositol polyphosphate-4-phosphatase type II B; inositol polyphosphate-4-phosphatase, type II; inositol polyphosphate-4-phosphatase, type II, 105kD; inositol polyphosphate-4-phosphatase, type II, 105kDa; Inpp4b; phosphatidylinositol-3,4-bisphosphate 4-phosphatase; phosphoinositide 4-phosphatase; type II inositol 3,4-bisphosphate 4-phosphatase; type II inositol-3,4-bisphosphate 4-phosphatase | |
| A synthesized peptide derived from human INPP4B (1-54AA). | |
| Primary | |
| -20°C | |
| INPP4B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title