missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INPP4B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558057
This item is not returnable.
View return policy
Description
Immunogen sequence: QDSIPHHSDY DEEEWDRVWA NVGKSLNCII AMVDKLIERD GGSEGSGGNN DGEKEPSLTD AIPSHPREDW YEQLYPLILT LKDCMGEVVN RAKQ Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 76%.
Catalyzes the hydrolysis of the 4-position phosphate of phosphatidylinositol 3,4-bisphosphate, inositol 1,3,4- trisphosphate and inositol 1,4-bisphosphate.
Specifications
| INPP4B | |
| Polyclonal | |
| Unconjugated | |
| INPP4B | |
| D-myo-inositol-3,4-bisphosphate 4-phosphohydrolase; E130107I17Rik; inositol polyphosphate 4-phosphatase II; 4-phosphatase II; inositol polyphosphate 4-phosphatase type II; inositol polyphosphate 4-phosphatase type II beta isoform; inositol polyphosphate-4-phosphatase type II 105kD; inositol polyphosphate-4-phosphatase type II B; inositol polyphosphate-4-phosphatase, type II; inositol polyphosphate-4-phosphatase, type II, 105kD; inositol polyphosphate-4-phosphatase, type II, 105kDa; Inpp4b; phosphatidylinositol-3,4-bisphosphate 4-phosphatase; phosphoinositide 4-phosphatase; type II inositol 3,4-bisphosphate 4-phosphatase; type II inositol-3,4-bisphosphate 4-phosphatase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8821 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15327 | |
| INPP4B | |
| Recombinant protein corresponding to Human INPP4B. Recombinant protein control fragment (Product #RP-96621). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction