Learn More
INPP1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12261-1-AP-20UL
This item is not returnable.
View return policy
Description
The INPP1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human INPP1. This antibody recognizes human antigen. The INPP1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| INPP1 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 3628 | |
| IgG |
| Western Blot | |
| 133 μg/mL | |
| Rabbit | |
| Primary | |
| Human |