missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INPP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557777
This item is not returnable.
View return policy
Description
Immunogen sequence: RELLCVSEKA ANIARACRQQ EALFQLLIEE KKEGEKNKKF AVDFKTLADV LVQEVIKQNM ENKFPGLEKN IFGEESNEFT N Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
This gene encodes the enzyme inositol polyphosphate-1-phosphatase, one of the enzymes involved in phosphatidylinositol signaling pathways. This enzyme removes the phosphate group at position 1 of the inositol ring from the polyphosphates inositol 1,4-bisphosphate and inositol 1,3,4-trisphophosphate.
Specifications
| INPP1 | |
| Polyclonal | |
| Unconjugated | |
| INPP1 | |
| 2300002C06Rik; AV137389; Inositol polyphosphate 1-phosphatase; inositol polyphosphate-1-phosphatase; Inpp1; IPP; IPPase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3628 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49441 | |
| INPP1 | |
| Recombinant protein corresponding to Human INPP1. Recombinant protein control fragment (Product #RP-96138). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction