missing translation for 'onlineSavingsMsg'
Learn More

Inositol Monophosphatase 3/IMPAD1 Antibody [HRP], Novus Biologicals™

Product Code. 30735103 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735103 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735103 Supplier Novus Biologicals Supplier No. AF7028H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Inositol Monophosphatase 3/IMPAD1 Polyclonal antibody specifically detects Inositol Monophosphatase 3/IMPAD1 in Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Inositol Monophosphatase 3/IMPAD1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias EC 3.1.3, EC 3.1.3.25, FLJ20421, IMP 3, IMPA3IMPase 3, inositol monophosphatase 3, inositol monophosphatase domain containing 1, Inositol monophosphatase domain-containing protein 1, Inositol-1(or 4)-monophosphatase 3, Myo-inositol monophosphatase A3
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Inositol Monophosphatase 3/IMPAD1, Asn145-His356, Accession # Q80V26
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 54928
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.