missing translation for 'onlineSavingsMsg'
Learn More

(-)-Inosine Dehydroxy-N-(6-iminohexyl)-2,2,2-trifluoroacetamide Phospho-morpholine, TRC

Product Code. 30397952
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397952 2.5 mg 2.5mg
30392674 10 mg 10mg
30395326 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397952 Supplier Toronto Research Chemicals Supplier No. TRCI6610152.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (-)-Inosine Dehydroxy-N-(6-iminohexyl)-2,2,2-trifluoroacetamide Phospho-morpholine
Analyte or Component Names (-)-Inosine Dehydroxy-N-(6-iminohexyl)-2,2,2-trifluoroacetamide Phospho-morpholine
Shipping Condition Room Temperature
Molecular Formula C12 H4 Br2 Cl2 O2
Synonym ((2R,3S,4R,5R)-3,4-Dihydroxy-5-(6-((6-(2,2,2-trifluoroacetamido)hexyl)amino)-9H-purin-9-yl)tetrahydrofuran-2-yl)methyl Morpholino Hydrogen Phosphate
InChI Formula InChI=1S/C12H4Br2Cl2O2/c13-5-1-9-10(2-6(5)14)18-12-4-8(16)7(15)3-11(12)17-9/h1-4H
Solution Type Neat
SMILES Clc1cc2Oc3cc(Br)c(Br)cc3Oc2cc1Cl
IUPAC Name 2,3-dibromo-7,8-dichlorodibenzo-p-dioxin
Industry Type Pharmaceutical
Molecular Weight (g/mol) 410.873
Quantity 2.5 mg
Formula Weight 407.796
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.