missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Inosine Dehydroxy-N-(6-iminohexyl)-2,2,2-trifluoroacetamide Phospho-morpholine, TRC
Specifications
Specifications
| Chemical Name or Material | (-)-Inosine Dehydroxy-N-(6-iminohexyl)-2,2,2-trifluoroacetamide Phospho-morpholine |
| Analyte or Component Names | (-)-Inosine Dehydroxy-N-(6-iminohexyl)-2,2,2-trifluoroacetamide Phospho-morpholine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H4 Br2 Cl2 O2 |
| Synonym | ((2R,3S,4R,5R)-3,4-Dihydroxy-5-(6-((6-(2,2,2-trifluoroacetamido)hexyl)amino)-9H-purin-9-yl)tetrahydrofuran-2-yl)methyl Morpholino Hydrogen Phosphate |
| InChI Formula | InChI=1S/C12H4Br2Cl2O2/c13-5-1-9-10(2-6(5)14)18-12-4-8(16)7(15)3-11(12)17-9/h1-4H |
| Solution Type | Neat |
| SMILES | Clc1cc2Oc3cc(Br)c(Br)cc3Oc2cc1Cl |
| IUPAC Name | 2,3-dibromo-7,8-dichlorodibenzo-p-dioxin |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?