missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Inosine Dehydroxy-N-(6-iminohexyl)-2,2,2-trifluoroacetamide Phosphate, TRC
Specifications
Specifications
| Chemical Name or Material | (-)-Inosine Dehydroxy-N-(6-iminohexyl)-2,2,2-trifluoroacetamide Phosphate |
| Analyte or Component Names | (-)-Inosine Dehydroxy-N-(6-iminohexyl)-2,2,2-trifluoroacetamide Phosphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H24 F17 N2 O2 P |
| Synonym | ((2R,3S,4R,5R)-3,4-Dihydroxy-5-(6-((6-(2,2,2-trifluoroacetamido)hexyl)amino)-9H-purin-9-yl)tetrahydrofuran-2-yl)methyl Dihydrogen Phosphate |
| InChI Formula | InChI=1S/C20H24F17N2O2P/c1-11(2)39(12(3)4)42(41-10-6-8-38)40-9-5-7-13(21,22)14(23,24)15(25,26)16(27,28)17(29,30)18(31,32)19(33,34)20(35,36)37/h11-12H,5-7,9-10H2,1-4H3 |
| Solution Type | Neat |
| SMILES | CC(C)N(C(C)C)P(OCCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)OCCC#N |
| IUPAC Name | 3-[[di(propan-2-yl)amino]-(4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-heptadecafluoroundecoxy)phosphanyl]oxypropanenitrile |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?