missing translation for 'onlineSavingsMsg'
Learn More

(-)-Inosine Dehydroxy-N-(6-iminohexyl)-2,2,2-trifluoroacetamide Phosphate, TRC

Product Code. 30390014
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390014 50 mg 50mg
30400780 5 mg 5mg
30401589 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390014 Supplier Toronto Research Chemicals Supplier No. TRCI66101050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (-)-Inosine Dehydroxy-N-(6-iminohexyl)-2,2,2-trifluoroacetamide Phosphate
Analyte or Component Names (-)-Inosine Dehydroxy-N-(6-iminohexyl)-2,2,2-trifluoroacetamide Phosphate
Shipping Condition Room Temperature
Molecular Formula C20 H24 F17 N2 O2 P
Synonym ((2R,3S,4R,5R)-3,4-Dihydroxy-5-(6-((6-(2,2,2-trifluoroacetamido)hexyl)amino)-9H-purin-9-yl)tetrahydrofuran-2-yl)methyl Dihydrogen Phosphate
InChI Formula InChI=1S/C20H24F17N2O2P/c1-11(2)39(12(3)4)42(41-10-6-8-38)40-9-5-7-13(21,22)14(23,24)15(25,26)16(27,28)17(29,30)18(31,32)19(33,34)20(35,36)37/h11-12H,5-7,9-10H2,1-4H3
Solution Type Neat
SMILES CC(C)N(C(C)C)P(OCCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)OCCC#N
IUPAC Name 3-[[di(propan-2-yl)amino]-(4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-heptadecafluoroundecoxy)phosphanyl]oxypropanenitrile
Industry Type Pharmaceutical
Molecular Weight (g/mol) 678.363
Quantity 50 mg
Formula Weight 678.13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.