missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Inosine 5'-monophosphate, disodium salt hydrate, 97%

Product Code. 10053523 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10053523 100 g Glass bottle 100g
10551281 5 g Glass bottle 5g
10236520 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10053523 Supplier Thermo Scientific Chemicals Supplier No. 226261000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 352195-40-5 | C10H11N4Na2O8P | 392.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352195-40-5
Molecular Formula C10H11N4Na2O8P
Molecular Weight (g/mol) 392.17
MDL Number MFCD00150372
InChI Key AANLCWYVVNBGEE-WCYUCLFNNA-L
Synonym inosine 5'-monophosphate disodium salt hydrate
PubChem CID 126963545
IUPAC Name disodium;[(2S,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl phosphate;hydrate
SMILES [Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1N=CNC2=O

Specifications

Melting Point 175.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 g
Beilstein 31, 26
Merck Index 14, 4977
Solubility Information Solubility in water: soluble
Spectrophotometry 96% min. (lambda max. 249nm,pH 7) molar absorptivity 12300+/-4%,on dry basis
Formula Weight 392.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Inosine 5'-monophosphate, disodium salt hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.