missing translation for 'onlineSavingsMsg'
Learn More

Inosine-5-diphosphoric Acid Disodium Salt, TRC

Product Code. 30390362
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390362 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390362 Supplier Toronto Research Chemicals Supplier No. TRCI6650131G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Inosine-5-diphosphoric Acid Disodium Salt
CAS 54735-61-4
Molecular Formula C10 H12 N4 Na2 O11 P2
Quantity 1 g
Synonym Inosine 5'-Diphosphate Disodium Salt
InChI Formula InChI=1S/C10H14N4O11P2.2Na/c15-6-4(1-23-27(21,22)25-26(18,19)20)24-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;;/h2-4,6-7,10,15-16H,1H2,(H,21,22)(H,11,12,17)(H2,18,19,20);;/q;2*+1/p-2/t4-,6-,7-,10-;;/m1../s1
SMILES O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)(O[Na])OP(=O)(O)O[Na])n2cnc3C(=O)N=CNc23
IUPAC Name [[(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methoxy-[hydroxy(sodiooxy)phosphoryl]oxyphosphoryl]oxysodium
Molecular Weight (g/mol) 472.149
Formula Weight 471.977
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.