missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inosine-5-diphosphoric Acid Disodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Inosine-5-diphosphoric Acid Disodium Salt |
| CAS | 54735-61-4 |
| Molecular Formula | C10 H12 N4 Na2 O11 P2 |
| Quantity | 5 g |
| Synonym | Inosine 5'-Diphosphate Disodium Salt |
| InChI Formula | InChI=1S/C10H14N4O11P2.2Na/c15-6-4(1-23-27(21,22)25-26(18,19)20)24-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;;/h2-4,6-7,10,15-16H,1H2,(H,21,22)(H,11,12,17)(H2,18,19,20);;/q;2*+1/p-2/t4-,6-,7-,10-;;/m1../s1 |
| SMILES | O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)(O[Na])OP(=O)(O)O[Na])n2cnc3C(=O)N=CNc23 |
| IUPAC Name | [[(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methoxy-[hydroxy(sodiooxy)phosphoryl]oxyphosphoryl]oxysodium |
| Molecular Weight (g/mol) | 472.149 |
| Formula Weight | 471.977 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?