Learn More
iNOS Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-18985-100UL
Description
The iNOS antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human iNOS. This antibody recognizes human, mouse, rat antigen. The iNOS antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
| iNOS | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite 488 | |
| 100 μL | |
| 4843 | |
| IgG |