missing translation for 'onlineSavingsMsg'
Learn More

iNOS Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18340143 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340143 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340143 Supplier Novus Biologicals Supplier No. NBP299091AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

iNOS Polyclonal specifically detects iNOS in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen iNOS
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias EC 1.14.13.39, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS type II, NOS2A, Peptidyl-cysteine S-nitrosylase NOS2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human iNOS.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cancer, HIF Target Genes, Hypoxia, Inflammation, Lipid and Metabolism, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 4843
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.