missing translation for 'onlineSavingsMsg'
Learn More

iNOS Antibody (4E5), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18430868 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430868 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430868 Supplier Novus Biologicals Supplier No. NBP222119AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

iNOS Monoclonal antibody specifically detects iNOS in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Intracellular)
TRUSTED_SUSTAINABILITY

Specifications

Antigen iNOS
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4E5
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias EC 1.14.13.39, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS type II, NOS2A, Peptidyl-cysteine S-nitrosylase NOS2
Host Species Mouse
Immunogen iNOS Antibody (4E5) was made to a purified recombinant fragment of human iNOS (C-terminus) expressed in E. coli. [UniProt# P35228]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cancer, HIF Target Genes, Hypoxia, Inflammation, Lipid and Metabolism, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 4843
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.