missing translation for 'onlineSavingsMsg'
Learn More

Inorganic Pyrophosphatase/PPA1 Antibody [PerCP], Novus Biologicals™

Product Code. 30733358 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733358 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733358 Supplier Novus Biologicals Supplier No. AF6557C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Inorganic Pyrophosphatase/PPA1 Polyclonal antibody specifically detects Inorganic Pyrophosphatase/PPA1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Inorganic Pyrophosphatase/PPA1
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias diphosphate phosphohydrolase, EC 3.6.1.1, inorganic diphosphatase, inorganic pyrophosphatase, inorganic pyrophosphatase 1, IOPPPMGC111556, PP1, PPase, PPcytosolic inorganic pyrophosphatase, pyrophosphatase (inorganic), pyrophosphatase (inorganic) 1, pyrophosphatase 1, Pyrophosphate phospho-hydrolase, SID6-8061
Host Species Sheep
Immunogen E. coli-derived recombinant human Inorganic Pyrophosphatase/PPA1, Met1-Asn289, Accession # Q15181
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5464
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.