missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inorganic Pyrophosphatase/PPA1 Antibody (4G4), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59471PE
This item is not returnable.
View return policy
Description
Inorganic Pyrophosphatase/PPA1 Monoclonal antibody specifically detects Inorganic Pyrophosphatase/PPA1 in Human samples. It is validated for Flow Cytometry
Specifications
| Inorganic Pyrophosphatase/PPA1 | |
| Monoclonal | |
| PE | |
| diphosphate phosphohydrolase, EC 3.6.1.1, inorganic diphosphatase, inorganic pyrophosphatase, inorganic pyrophosphatase 1, IOPPPMGC111556, PP1, PPase, PPcytosolic inorganic pyrophosphatase, pyrophosphatase (inorganic), pyrophosphatase (inorganic) 1, pyrophosphatase 1, Pyrophosphate phospho-hydrolase, SID6-8061 | |
| Recombinant human Inorganic Pyrophosphatase/PPA1 (1-289aa) purified from E. coli | |
| 0.1 mL | |
| metabolism, Signal Transduction | |
| 5464 | |
| Store at 4°C in the dark. | |
| IgG2b λ |
| Flow Cytometry | |
| 4G4 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction