missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ inoLab™ pH 7310P Set 2 with Stand
Click to view available options
Range (pH):
-2 to 20
Unit Size:
Each
Specifications
Specifications
| Includes | 3 in 1 pH electrode, meter with universal power supply, electrode arm and operation manual. 3 in 1 pH electrode SenTix™ 41, buffers, 3mol/L KCl, software and USB cable, integrated thermal printer |
| Accuracy (Temperature) | ±0.1K |
| Accuracy (mV) | ±0.3, ±1 |
| Accuracy (pH) | ±0.005, ±0.01 |
| Data Management Details | Up to 10 records calibration memory, 500 manual datasets/5000 automatic memory |
| For Use With (Application) | For measurements/documentation according GLP/AQA, AC and battery operation |
| Range (pH) | -2 to 20 |
| Resolution (pH) | -2 to 20 ±0.01, -2 to 19.999 ±0.005 |
| Range (mV) | ±1200, ±0.3 |
| Resolution (mV) | ±(1200 ±1)mV |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction