missing translation for 'onlineSavingsMsg'
Learn More

Inobrodib, MedChemExpress

Product Code. 16474528
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474528 10 mg 10mg
16420837 50 mg 50mg
16407247 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16474528 Supplier Medchem Express Supplier No. HY111784

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Inobrodib (CCS1477) is an orally active, potent, and selective inhibitor of the p300/CBP bromodomain. Inobrodib binds to p300 and CBP with Kd values of 1.3 and 1.7 nM, respectively, and with 170/130-fold selectivity compared with BRD4 with a Kd of 222 nM. CCS1477 inhibits cell proliferation in prostate cancer cell lines and decreases androgen receptor (AR)- and C-MYC-regulated gene expression.

Inobrodib (CCS1477) is an orally active, potent, and selective inhibitor of the p300/CBP bromodomain. Inobrodib binds to p300 and CBP with Kd values of 1.3 and 1.7 nM, respectively, and with 170/130-fold selectivity compared with BRD4 with a Kd of 222 nM. CCS1477 inhibits cell proliferation in prostate cancer cell lines and decreases androgen receptor (AR)- and C-MYC-regulated gene expression.

Specifications

Chemical Name or Material Inobrodib
CAS 2222941-37-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H32F2N4O3
Quantity 10 mg
Synonym CCS1477
Solubility Information DMSO : 100 mg/mL (187.06 mM; Need ultrasonic)
SMILES O=C1N(C2=CC=C(F)C(F)=C2)[C@H](C3=NC4=CC(C5=C(C)ON=C5C)=CC=C4N3[C@H]6CC[C@H](OC)CC6)CCC1
Molecular Weight (g/mol) 534.6
Formula Weight 534.6
Percent Purity 99.41%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.