missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INO80 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78758AF488
This item is not returnable.
View return policy
Description
INO80 Polyclonal antibody specifically detects INO80 in Human samples. It is validated for Western Blot
Specifications
| INO80 | |
| Polyclonal | |
| Western Blot | |
| EC 3.6.1, EC 3.6.1.23, EC 3.6.4.12, hINO80INO80 complex subunit A, homolog of yeast INO80, Ino80, INO80 complex homolog 1 (S. cerevisiae), INO80 homolog (S. cerevisiae), INO80A, INOC1putative DNA helicase INO80 complex homolog 1, KIAA1259 | |
| The immunogen for this product maps to a region between residue 1400 and 1450 of human INO80 homolog using the numbering given in entry NP_060023.1 (GeneID 54617). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 54617 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction