missing translation for 'onlineSavingsMsg'
Learn More

INO5042, MedChemExpress

Product Code. 16403149
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16403149 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16403149 Supplier Medchem Express Supplier No. HYU00094
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress INO5042, a thiazole fused 1,4-naphthoquinone compound, and exhibits anti-inflammation activity.

INO5042, a thiazole fused 1,4-naphthoquinone compound, and exhibits anti-inflammation activity.

Specifications

Chemical Name or Material INO5042
CAS 14782-19-5
Color Orange
Purity Grade Notes Research
Molecular Formula C15H7NO3S
Quantity 1 mg
Solubility Information DMSO : 2 mg/mL (7.11 mM; ultrasonic and adjust pH to 3 with HCl)
SMILES O=C(C1=C2SC(C3=CC=CO3)=N1)C4=C(C=CC=C4)C2=O
Molecular Weight (g/mol) 281.29
Formula Weight 281.29
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.