missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ 40nm Colloidal Gold Nanoparticles 10 OD

Product Code. 18706973 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
100mL
10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18706973 10 mL 10mL
18765073 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only
Product Code. 18706973 Supplier Novus Biologicals™ Supplier No. 2100010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
For Research Use Only
  • Uniform Shape
  • Low co-efficient of variation in terms of size
  • High efficiency antibody binding for maximum sensitivity
  • Available at high concentrations to simplify assay development
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C. Do not freeze.
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.