missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ InnovaCoat (R) GOLD - 80nm Goat Anti-Rabbit IgG Gold (10OD)

Product Code. p-7111307 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
1 mL
Unit Size:
0.2mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18035576 0.2 mL 0.2mL
18046784 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18035576 Supplier Novus Biologicals™ Supplier No. 2210200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Unique coated nanoparticle with a strongly bound protective surface coat

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C. Do not freeze.
Quantity 0.2 mL
Target InnovaCoat GOLD - 80nm Goat Anti-Rabbit IgG Gold (10OD)
Product Type Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.