missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ InnovaCoat (R) GOLD 10nm Mini Antibody Labeling Kit

Product Code. 18767023 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 Tests
3 Tests
Unit Size:
10 tests
3 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18767023 10 Tests 10 tests
18044414 3 Tests 3 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only
Product Code. 18767023 Supplier Novus Biologicals™ Supplier No. 2280010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only

Use simple pipetting operations to covalently attach antibodies, proteins and other biomolecules to ultra-stable InnovaCoat GOLD nanoparticles. Hands-on time is about two minutes and the conjugate is ready within fifteen.

Features
  • Proprietary surface coat reduces risk of sub-optimal antibody performance
  • Ultra-stable
  • Covalently link antibodies and analytes
  • Simple conjugation processes
  • Different surface chemistries
Benefits
  • Antibody-GOLD conjugates in just 15 minutes
  • Enhanced assay sensitivity
  • Protective coat prevents metal-antibody interactions
  • Coating procedure not dependent on isoelectric point of antibody
  • Rapid screening of multiple antibodies for assay development
  • Reduced antibody loading without aggregation
  • No pH titrations
  • Fully scalable – from R&D to production
  • Custom InnovaCoat™ labeling services
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C. Do not freeze.
Quantity 10 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.