missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INMT Recombinant Rabbit Monoclonal Antibody (23GB2160), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552478
This item is not returnable.
View return policy
Description
N-methylation of endogenous and xenobiotic compounds is a major method by which they are degraded. This gene encodes an enzyme that N-methylates indoles such as tryptamine.
Specifications
| INMT | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95050, P40936 | |
| INMT | |
| A synthesized peptide derived from human TEMT (1-150AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2160 | |
| Unconjugated | |
| INMT | |
| Amine N-methyltransferase; aromatic alkylamine N-methyltransferase; arylamine N-methyltransferase; indolamine N-methyltransferase; Indolethylamine N-methyltransferase; Inmt; nicotine N-methyltransferase; Temt; Thioether S-methyltransferase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11185, 21743, 368066 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction