missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INMT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21578-1-AP-150UL
This item is not returnable.
View return policy
Description
N-methylation of endogenous and xenobiotic compounds is a major method by which they are degraded. This gene encodes an enzyme that N-methylates indoles such as tryptamine.Specifications
| INMT | |
| Polyclonal | |
| Unconjugated | |
| INMT | |
| Amine N methyltransferase, Arylamine N methyltransferase, Indolamine N methyltransferase, INMT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11185, 21743 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95050, P40936 | |
| Inmt | |
| INMT Fusion Protein Ag16121 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction