missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INMT Polyclonal Antibody
GREENER_CHOICE

Product Code. 15959084
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15959084 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15959084 Supplier Invitrogen™ Supplier No. PA566379

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RVLKCDVHLGN PLAPAVLPLA DCVLTLLAME CACCSLDAYR AALCNLASLL Highest antigen sequence identity to the following orthologs - mouse 67%, rat 67%.

N-methylation of endogenous and xenobiotic compounds is a major method by which they are degraded. This gene encodes an enzyme that N-methylates indoles such as tryptamine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INMT
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INMT
Gene Accession No. O95050
Gene Alias Amine N-methyltransferase; aromatic alkylamine N-methyltransferase; arylamine N-methyltransferase; indolamine N-methyltransferase; Indolethylamine N-methyltransferase; Inmt; nicotine N-methyltransferase; Temt; Thioether S-methyltransferase
Gene Symbols INMT
Host Species Rabbit
Immunogen Recombinant Human INMT. Recombinant protein control fragment (Product #RP-107054).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11185
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.