missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INMT Antibody (31G7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26197-100ul
This item is not returnable.
View return policy
Description
INMT Monoclonal antibody specifically detects INMT in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| INMT | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| 31G7 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| Amine N-methyltransferase, Aromatic alkylamine N-methyltransferase, Arylamine N-methyltransferase, EC 2.1.1, EC 2.1.1.49, Indolamine N-methyltransferase, indolethylamine N-methyltransferase, MGC125940, MGC125941, nicotine N-methyltransferase | |
| A synthesized peptide derived from Human INMT [UniProt O95050] | |
| 100 μL | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 11185 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction