missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Inlet Liners for Split Injection

Product Code. 13194382
Encompass_Preferred
Change view
Click to view available options
Deactivation:
Base Deactivated
Siltek
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Deactivation unitSize
13194382 Siltek -
13174382 Base Deactivated Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13194382 Supplier Restek™ Supplier No. 21023214.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fully deactivated to prevent degradation or adsorption of active compounds

  • Designed with mixing chambers and tortuous flow paths to fully vaporize samples into a homogenous vapor cloud before reaching split point
  • Wool packing traps nonvolatile residue and prevents column contamination when analyzing dirty samples
  • Liners with Siltek™-deactivated surface offer high temperature stability, better durability and low bleed
  • O.D. × L of all liners: 6.3 × 78.5mm; I.D. 4mm

Split liners with wool

  • Provide a large surface area to deliver a uniform vapor cloud to the split point
  • Economical and reproducible performance
  • Compatible with Agilent™ 7673 autosampler
Precision™ liners
  • Offer high reproducibility and maximum vaporization
  • Wool placed at the injection point helps wipe the needle during injection and stays in position during pressure pulses and injection

†To order pack quantities, contact your Customer Service Representative.

Specifications

Deactivation Siltek
Geometry Precision
Quantity 5/Pk.
Length (Metric) 7.85 cm
Packing Material Deactivated Wool
Product Type Inlet Liner
Diameter (Metric) Inner 4.0 mm
Diameter (Metric) Outer 6.3 mm
For Use With (Equipment) Agilent GCs Equipped with Split/Splitless Inlets
Liner Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.