missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Reach pH Electrodes

Product Code. 10523375 Shop All Mettler Toledo Products
Change view
Click to view available options
Connector Types:
Multi-Pin
S7
Includes:
Electrode only
Temperature Probe (NTC 30 kΩ)
Temperature Probe (Pt-1000)
Length (Metric) Stem:
225 mm
425 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Includes Length (Metric) Stem unitSize
10523375 Multi-Pin Temperature Probe (Pt-1000) 425 mm Each
10049392 Multi-Pin Temperature Probe (NTC 30 kΩ) 425 mm Each
15529601 S7 Electrode only 225 mm Each
15569601 S7 Electrode only 425 mm Each
15579601 Multi-Pin Temperature Probe (NTC 30 kΩ) 225 mm Each
15599601 Multi-Pin Temperature Probe (Pt-1000) 225 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10523375 Supplier METTLER TOLEDO™ Supplier No. 51343062
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A pH electrode with an integrated temperature sensor featuring an extended shaft for measuring a wide range of samples in large vessels.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Pilot reactor
  • Tank and Barrel
  • Large samples
  • Deep vessels

Accessories & Care

  • Ceramic junction
  • HA glass membrane
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Description pH electrode InLab Reach Pt1000-425
Connector Types Multi-Pin
Junction Ceramic
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode
Filling Solution Type 3mol/L KCl
For Use With (Application) Accurate measurement in a wide range of sample types
For Use With (Equipment) Multi-Pin to BNC (30281896)
Includes Temperature Probe (Pt-1000)
Internal Reference ARGENTHAL with Ag+ trap
Membrane HA Glass
Model InLab Reach Pt1000-425
Probe PT1000
Response Time <20 sec.
Storage Requirements 3mol/L KCl
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 15.7 in.
Length (Metric) 400 mm
Length (Metric) Stem 425 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.