missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Pure pH Electrodes

Product Code. 11384213
Change view
Click to view available options
Connector Types:
Multi-Pin
S7
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types unitSize
11384213 Multi-Pin Each
15559601 S7 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384213 Supplier METTLER TOLEDO™ Supplier No. 51344172
  • ONLINE EXCLUSIVE PRICE
    £698.29
    £570.98 / Pack of 1
    Save £127.31
    18% Off
View all promotions
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

A glass pH electrode designed for ion-weak-samples such as ultrapure water and deionized water.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Pure water
  • Ultrapure water
  • Deionized water
  • Waste water
  • Highly saline solution, sea water
  • Low-ion strength

Accessories & Care

  • Immovable glass sleeve junction
  • LoT membrane glass
  • InLab Pure: refillable FRISCOLYT-B™ (51343185) - keep probe filled with electrolyte for accurate results
  • InLab Pure Pro-ISM: refillable 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Description pH electrode InLab Pure Pro-ISM
Connector Types Multi-Pin
Junction Immovable Glass Sleeve
Material Glass
pH Range 1 to 11
Temperature (Metric) Operating 0°C to 80°C
Type Combination Electrode
Filling Solution Type 3 mol/L KCl Reference, 1mol/L KCl Bridge
For Use With (Application) Measuring samples at low temperatures or with low ion concentrations
For Use With (Equipment) Multi-Pin to BNC/RCA (30281896)
Includes ISM Temperature Probe
Internal Reference ARGENTHAL with Ag+ trap
Membrane LoT Glass
Model InLab™ Pure Pro-ISM
Probe NTC 30 kω
Storage Requirements 3mol/L KCl
Temperature (English) Operating 32°F to 176°F
Temperature (Metric) Range 0°C to 80°C
Length (English) 6.7 in.
Length (Metric) 170 mm
Diameter (English) 0.47 in.
Diameter (Metric) Body 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.