Learn More
Mettler Toledo™ InLab™ OptiOx Pro-DES™ DO Sensing Cap
Shop All Mettler Toledo Products
Click to view available options
Type:
RDO Fast Cap Replacement Kit
Description
- Optical sensing cap compatible with InLab OptiOx Pro-DES sensor, which can be purchased separately for replacement; it has a lifespan of 18 months, ensuring long-term usability and cost-effective maintenance
Specifications
Specifications
| Type | RDO Fast Cap Replacement Kit |