Learn More
Mettler Toledo™ InLab™ OptiOx Pro-DES™ DO BOD Adapter
GREENER_CHOICE
Product Code. 31454068 Shop All Mettler Toledo Products
Click to view available options
Type:
BOD Adapter
Product Code. 31454068
Brand: Mettler Toledo™ 30786584
Enables easy installation of the sensor for use in common BOD applications.
  • Use the BOD adapter to install the InLab OptiOx Pro-DES sensor on all common BOD applications.
TRUSTED_SUSTAINABILITY
Specifications
Type BOD Adapter